Order Entry
Sweden
ContactUsLinkComponent
 
Anti-ATP6V1E1 Mouse Monoclonal Antibody [clone: 4E11]
  ABNOH00000529-M02
 :  Abnova
undefined
Anti-ATP6V1E1 Mouse Monoclonal Antibody [clone: 4E11]
  ABNOH00000529-M02
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1
  • Antigen symbol:
    ATP6V1E1
  • Clonality:
    Monoclonal
  • Clone:
    4E11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    529
  • Antigen synonyms:
    ATP6V1E|Vma4|ATP6E2|ATP6E|P31
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ATP6V1E1
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human