Anti-ATP6V1E1 Rabbit Polyclonal Antibody
ABNOH00000529-D01P
: Abnova
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1
- Antigen symbol:ATP6V1E1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:529
- Antigen synonyms:ATP6V1E|Vma4|ATP6E2|ATP6E|P31
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ATP6V1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse