Anti-ACVR1 Mouse Monoclonal Antibody [clone: 2D5]
Catalog # ABNOH00000090-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Activin A Receptor, Type 1
- Antigen symbol:ACVR1
- Clonality:Monoclonal
- Clone:2D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:90
- Antigen synonyms:ACTRI|ALK2|ACVR1A|TSRI|FOP|SKR1|ACVRLK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ACVR1
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human