Anti-EGR2 Mouse Monoclonal Antibody [clone: OTI1B12]
Catalog # ORIGTA505382
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:early growth response 2
- Antigen symbol:EGR2
- Clonality:Monoclonal
- Clone:OTI1B12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:none
- Form:Liquid
- Amino acid number:156 to 476
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:50.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100
Type: Primary
Antigen: EGR2 (early growth response 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human