Anti-CAPN2 Mouse Monoclonal Antibody [clone: OTI3G1]
Catalog # ORIGTA504280
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CAPN2
- Clonality:Monoclonal
- Clone:OTI3G1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:79.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: CAPN2 (calpain 2, (m/II) large subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: