Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI3B12]
ORIGTA501527
: OriGene
- Antibody type:Primary
- Antigen name:butyrophilin, subfamily 1, member A1
- Antigen symbol:BTN1A1
- Clonality:Monoclonal
- Clone:OTI3B12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:58.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
- Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: BTN1A1 (butyrophilin, subfamily 1, member A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human