Anti-OR8U1 Rabbit Polyclonal Antibody
Catalog # ORIGTA316226
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 8, subfamily U, member 1
- Antigen symbol:OR8U1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000
Type: Primary
Antigen: OR8U1 (olfactory receptor, family 8, subfamily U, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human