Order Entry
Sweden
ContactUsLinkComponent
Anti-FSD1 Goat Antibody
Anti-FSD1 Goat Antibody
Catalog # ORIGTA311519
Supplier:  OriGene
Anti-FSD1 Goat Antibody
Catalog # ORIGTA311519
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibronectin type III and SPRY domain containing 1
  • Antigen symbol:
    FSD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.Aliquot and store at -20C. Minimize freezing and thawing.
  • Sequence:
    C-KVQDIKAREKDGKGR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: FSD1 (fibronectin type III and SPRY domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human