Anti-AIMP1 Goat Antibody
Catalog # ORIGTA311310
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
- Antigen symbol:AIMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.Aliquot and store at -20C. Minimize freezing and thawing.
- Sequence:NNDAVLKRLEQK-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.3-1?g/ml
Type: Primary
Antigen: AIMP1 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat