Order Entry
Sweden
ContactUsLinkComponent
Anti-PTCH1 Rabbit Polyclonal Antibody
Anti-PTCH1 Rabbit Polyclonal Antibody
Catalog # ORIGTA309821
Supplier:  OriGene
Anti-PTCH1 Rabbit Polyclonal Antibody
Catalog # ORIGTA309821
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    patched 1
  • Antigen symbol:
    PTCH1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS and 0,02% Sodium Azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 μL
  • Pk:
    0,1 ml

Specifications

About this item

Recommended Dilutions:

Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: