Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27b]
Catalog # ORIGTA309765
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Neurofibromin 1
- Antigen symbol:NF1
- Clonality:Monoclonal
- Clone:McNFn27b
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Guinea pig,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:27 to 41
- Storage buffer:PBS and 0,05% Sodium Azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:200 μL
- Pk:0,2 ml
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: NF1 (neurofibromin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human, Mouse, Rat