Order Entry
Sweden
ContactUsLinkComponent
Anti-NAPSA Mouse Monoclonal Antibody [clone: TMU-Ad02]
Anti-NAPSA Mouse Monoclonal Antibody [clone: TMU-Ad02]
Catalog # ORIGTA309758
Supplier:  OriGene
Anti-NAPSA Mouse Monoclonal Antibody [clone: TMU-Ad02]
Catalog # ORIGTA309758
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Napsin A
  • Antigen symbol:
    NAPSA
  • Clonality:
    Monoclonal
  • Clone:
    TMU-Ad02
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Tris-glycine, 150mM NaCl pH 7,5 and 0,05% Sodium Azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 μL
  • Pk:
    0,1 ml

Specifications

About this item

Recommended Dilutions:

Type: Primary
Antigen: NAPSA (napsin A aspartic peptidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human