Anti-MLXIPL Rabbit Polyclonal Antibody
Catalog # ORIGTA309750
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:MLX interacting Protein-like
- Antigen symbol:MLXIPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:800 to 852
- Storage buffer:in Tris-citrate/phosphate, pH 7-8 and 0,01% Sodium Azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μL
- Pk:0,1 ml
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: MLXIPL (MLX interacting protein-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat