Anti-LRRK2 Rabbit Polyclonal Antibody
Catalog # ORIGTA309736
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeat kinase 2
- Antigen symbol:LRRK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:2500 to 2527
- Storage buffer:Tris-citrate/phosphate, pH 7-8 and 0,1% Sodium Azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μL
- Pk:0,1 ml
Specifications
About this item
- Recommended Dilutions:
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: