Anti-MAP2K1 Rabbit Antibody
Catalog # ORIGTA309167
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 1
- Antigen symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Modification:phospho-Thr292
- Storage buffer:100 µl in 10 mM HEPES (pH 7,5), 150 mM NaCl, 100 µg per ml BSA and 50% glycerol.
- Molecular weight:45 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: