Anti-GAPDHS Mouse Antibody [clone: 1D4]
Katalog # ORIGTA309157
Leverantör: OriGene
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
- Antigensymbol:GAPDHS
- Klonalitet:Polyclonal
- Klon:1D4
- Konjugation:Unconjugated
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgM
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Korsad adsorption:none
- Mått:concentrated culture supernatant
- Form:Liquid
- Storage buffer:total IgM fraction + 10 mM NaN3.
- Molekylvikt:38 kDa
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 µl
- Förp.:100 µG
Specifikationer
Om denna produkt
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100
Type: Primary
Antigen: GAPDHS (glyceraldehyde-3-phosphate dehydrogenase, spermatogenic)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat