Order Entry
Sweden
ContactUsLinkComponent
Anti-Myc Tag None Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9E10]
Anti-Myc Tag None Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9E10]
  ORIGTA150054
 :  OriGene
Anti-Myc Tag None Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 9E10]
  ORIGTA150054
 :  OriGene

 

  • Antibody type:
    Tag
  • Antigen name:
    Myc tag
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Clone:
    9E10
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

 

 

Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: None
Isotype: none
Reactivity: