Anti-DDK Tag Antibody Agarose beads
Catalog # ORIGTA150037
Supplier: OriGene
Specifications
- Antibody type:Tag
- Antigen name:DDK Tag
- Antigen symbol:DDK
- Conjugation:Agarose
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:1 ml
Specifications
About this item
Type: Tag
Antigen: DDK Tag
Clonality:
Clone:
Conjugation: Agarose beads
Epitope:
Host: None
Isotype: none
Reactivity: