Om denna produkt
The HIB19 antibody reacts with human CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21, CD81 and CD225, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.
The HIB19 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Specifikationer
- Catalog No:
- 60-0199-T100
- 60-0199-T025
- Antigen symbol:
- CD19
- CD19
- Antigen name:
- CD19 molecule
- CD19 molecule
- Antigen synonyms:
- B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
- B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- HIB19
- HIB19
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 930
- 930
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice