Order Entry
Sweden
ContactUsLinkComponent
Anti-EMR1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: BM8.1]
Anti-EMR1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: BM8.1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-EMR1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: BM8.1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-4801-U100
  • 35-4801-U500
  • 35-4801-U025
  • Antigen symbol:
  • ADGRE1
  • ADGRE1
  • ADGRE1
  • Antigen name:
  • Adhesion G Protein-Coupled Receptor E1
  • Adhesion G Protein-Coupled Receptor E1
  • Adhesion G Protein-Coupled Receptor E1
  • Antigen synonyms:
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • BM8.1
  • BM8.1
  • BM8.1
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 13733
  • 13733
  • 13733
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information