Order Entry
Sweden
ContactUsLinkComponent
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-0081-U025
  • 35-0081-U100
  • 35-0081-U500
  • Antigen symbol:
  • CD8A
  • CD8A
  • CD8A
  • Antigen name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 53-6.7
  • 53-6.7
  • 53-6.7
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 12525
  • 12525
  • 12525
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information