Order Entry
Sweden
ContactUsLinkComponent
Anti-FUT3 Rabbit Polyclonal Antibody
Anti-FUT3 Rabbit Polyclonal Antibody
Catalog # ABGEAP9410B
Leverantör:  Abgent
undefined
Anti-FUT3 Rabbit Polyclonal Antibody
Catalog # ABGEAP9410B
Leverantör:  Abgent

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    Fucosyltransferase 3
  • Antigen symbol:
    FUT3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Gene ID:
    83984
  • Antigen synonyms:
    CD174|FT3B|Les|LE|FucT-III
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    GLPRRQKRGVLYPDC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse Tssk6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 204-232 amino acids from the C-terminal region of mouse Tssk6.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifikationer

Om denna produkt

Type: Primary
Antigen: FUT3 (fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human