Order Entry
Sweden
ContactUsLinkComponent
Anti-MAGI1 Rabbit Polyclonal Antibody
Anti-MAGI1 Rabbit Polyclonal Antibody
Catalog # ABGEAP8173A
Supplier:  Abgent
undefined
Anti-MAGI1 Rabbit Polyclonal Antibody
Catalog # ABGEAP8173A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Membrane associated guanylate kinase, WW and PDZ domain containing 1
  • Antigen symbol:
    MAGI1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Gene ID:
    7025
  • Antigen synonyms:
    Magi1d|AIP-3|BAIAP1|BAP-1|AIP3|BAP1|WWP3|MAGI-1|TNRC19
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    TPGTAGDKGQGPPGC
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This NR2F1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 48-76 amino acids from the N-terminal region of human NR2F1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAGI1 (membrane associated guanylate kinase, WW and PDZ domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human