Order Entry
Sweden
ContactUsLinkComponent
Anti-CYP17A1 Rabbit Polyclonal Antibody
Anti-CYP17A1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7879C
Supplier:  Abgent
undefined
Anti-CYP17A1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7879C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome P450, Family 17, Subfamily A, Polypeptide 1
  • Antigen symbol:
    CYP17A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    steroid 17-alpha-monooxygenase,17?-hydroxylase/17,20 lyase/17,20 desmolase|CYP17|S17AH|P450C17|CPT7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CYP17A1 (cytochrome P450, family 17, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human