Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Anti-PTRF Rabbit Polyclonal Antibody

  ABGEAP7421B
 :   Abgent
Anti-PTRF Rabbit Polyclonal Antibody
  ABGEAP7421B
 :  Abgent
Anti-PTRF Rabbit Polyclonal Antibody
Anti-PTRF Rabbit Polyclonal Antibody
 
4 075,00 kr
   
1 * 400 µl
 

Type: Primary
Antigen: PTRF (polymerase I and transcript release factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human

 
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • Antibody type:
    Primary
  • Epitope:
    C-terminal
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Shipping temperature:
    Ice
  • Antigen name:
    Polymerase I and transcript release factor
  • Cat. No.:
    ABGEAP7421B
  • Reactivity:
    Human
  • Antigen synonyms:
    CAVIN|cavin-1|FKSG13|Polymerase I and transcript release factor|CAVIN1|PTRF|Cavin-1|CGL4
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    PTRF
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.