Anti-CA5B Rabbit Polyclonal Antibody
Catalog # ABGEAP7308B
Leverantör: Abgent
Specifikationer
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 5B, mitochondrial
- Antigen symbol:CA5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:CA-VB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: CA5B (carbonic anhydrase VB, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse