Order Entry
Sweden
ContactUsLinkComponent
 
Anti-WNT4 Rabbit Polyclonal Antibody
  ABGEAP6683C
 :  Abgent
undefined
Anti-WNT4 Rabbit Polyclonal Antibody
  ABGEAP6683C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Wingless-type MMTV integration site family, member 4
  • Antigen symbol:
    WNT4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    WNT-4|SERKAL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse