Order Entry
Sweden
ContactUsLinkComponent
Anti-CAV3 Rabbit Polyclonal Antibody
Anti-CAV3 Rabbit Polyclonal Antibody
Catalog # ABGEAP6516A
Supplier:  Abgent
undefined
Anti-CAV3 Rabbit Polyclonal Antibody
Catalog # ABGEAP6516A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caveolin 3
  • Antigen symbol:
    CAV3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    M-caveolin|VIP21|LGMD1C|MGC126129|VIP-21|caveolin 3|MGC126100|Caveolin-3|CAV3|MGC126101|LQT9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CAV3 (caveolin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human