Anti-ZBTB7B Rabbit Polyclonal Antibody
Catalog # ABGEAP6370C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger And BTB Domain Containing 7B
- Antigen symbol:ZBTB7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:THPOK|ZNF857B|ZBTB15|c-KROX|ZFP67|hcKROX|ZFP-67|CKROX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZBTB7B (zinc finger and BTB domain containing 7B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse