Anti-ELK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6282A
Leverantör: Abgent
Specifikationer
- Antibody type:Primary
- Antigen name:ELK1, ETS transcription factor
- Antigen symbol:ELK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:PRO0593|EC 2.7.11.1|HHIPK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human