Anti-FOXO4 Rabbit Polyclonal Antibody
Catalog # ABGEAP6193A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box O4
- Antigen symbol:FOXO4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Antigen synonyms:MLLT7|AFX1|AFX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: FOXO4 (forkhead box O4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human