Anti-TCF4 Rabbit Polyclonal Antibody
Catalog # ABGEAP5756A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor 4
- Antigen symbol:TCF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:TCF7L2|TCF4|TCF-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TCF4 (transcription factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human