Order Entry
Sweden
ContactUsLinkComponent
 
Anti-SWSAP1 Rabbit Polyclonal Antibody
  ABGEAP5547C
 :  Abgent
undefined
Anti-SWSAP1 Rabbit Polyclonal Antibody
  ABGEAP5547C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Core-binding Factor, Beta subunit
  • Antigen symbol:
    C19orf39
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    Pebpb2|PEA2|Pebp2|AI893578|PEBP2b
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SWSAP1 (SWIM-type zinc finger 7 associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse