Anti-CNNM4 Rabbit Polyclonal Antibody
ABGEAP5419C
: Abgent
- Antibody type:Primary
- Antigen name:Granzyme M
- Antigen symbol:CNNM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:MET1|LMET1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CNNM4 (cyclin M4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human