Order Entry
Sweden
ContactUsLinkComponent
Anti-CD300LB Rabbit Polyclonal Antibody
Anti-CD300LB Rabbit Polyclonal Antibody
Catalog # ABGEAP5203A
Supplier:  Abgent
undefined
Anti-CD300LB Rabbit Polyclonal Antibody
Catalog # ABGEAP5203A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD300 Molecule-Like Family Member B
  • Antigen symbol:
    CD300LB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    IREM-3|CD300b|TREM5|CLM7|CLM-7|CMRF35-A2|IREM3|TREM-5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CD300LB (CD300 molecule-like family member b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human