Order Entry
Sweden
ContactUsLinkComponent
 
Anti-HVCN1 Rabbit Polyclonal Antibody
  ABGEAP5042A
 :  Abgent
undefined
Anti-HVCN1 Rabbit Polyclonal Antibody
  ABGEAP5042A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Hydrogen Voltage-Gated Channel 1
  • Antigen symbol:
    HVCN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Hydrogen voltage-gated channel 1|HVCN1|HV1|Voltage-gated hydrogen channel 1|VSOP|UNQ578/PRO1140
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: HVCN1 (hydrogen voltage-gated channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human