Anti-SRSF2 Rabbit Polyclonal Antibody
Catalog # ABGEAP2800C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Splicing Factor Serine/arginine-rich 2
- Antigen symbol:SFRS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Splicing factor|SC-35|SFRS2|35 kDa|Splicing factor SC35|Protein PR264|arginine/serine-rich 2|Splicing component
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SRSF2 (serine/arginine-rich splicing factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human