Anti-MOGS Rabbit Polyclonal Antibody
Catalog # ABGEAP2315B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Mannosyl-Oligosaccharide Glucosidase
- Antigen symbol:MOGS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:CDG2B|GCS1|CWH41|DER7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MOGS (mannosyl-oligosaccharide glucosidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse