Order Entry
Sweden
ContactUsLinkComponent
Anti-PSMD4 Rabbit Polyclonal Antibody
Anti-PSMD4 Rabbit Polyclonal Antibody
Catalog # ABGEAP2103B
Supplier:  Abgent
undefined
Anti-PSMD4 Rabbit Polyclonal Antibody
Catalog # ABGEAP2103B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome (prosome, macropain) 26S Subunit, non-ATPase 4
  • Antigen symbol:
    PSMD4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    AF|ASF|S5A|Rpn10|pUB-R5|26S proteasome regulatory subunit S5A|Multiubiquitin chain binding protein|MCB1|26S proteasome non-ATPase regulatory subunit 4|AF-1|Antisecretory factor-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PSMD4 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human