Anti-KCNJ15 Rabbit Polyclonal Antibody
Catalog # ABGEAP17113B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Potassium Inwardly-rectifying Channel Subfamily J Member 15
- Antigen symbol:KCNJ15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:3772
- Antigen synonyms:IRKK|KIR1.3|KIR4.2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:42.577 kDa
- Sequence:KYRQEDQRERELRTC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KCNJ15 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 339-367 amino acids from the C-terminal region of human KCNJ15.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: KCNJ15 (potassium inwardly-rectifying channel, subfamily J, member 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human