Anti-MARK1 Rabbit Polyclonal Antibody
ABGEAP14269B
: Abgent
- Antibody type:Primary
- Antigen name:MAP/microtubule affinity-regulating kinase 1
- Antigen symbol:MARK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:226778
- Antigen synonyms:Serine/threonine-protein kinase p78|ELKL motif kinase 1|Par1c|EMK-2|CTAK1|EMK-1|PAR1 homolog c|cTAK1|MARK3|MARK|Serine/threonine-protein kinase MARK1|MARK4|PAR1 homolog|Par1b|Protein kinase STK10|MAP/microtubule affinity-regulating kinase 4|PAR1 homolog b|MAP/microtubule affinity-regulating kinase 3|Ser/Thr protein kinase PAR-1|MARKL1|KIAA1860|C-TAK1|EMK1|Par-1a|MARK1|EMK2|MAP/microtubule affinity-regulating kinase 1|Serine/threonine-protein kinase MARK2|MAP/microtubule affinity-regulating kinase 2|Cdc25C-associated protein kinase 1|KIAA1477|Par-1c|MARK2|MAP/microtubule affinity-regulating kinase-like 1|ELKL motif kinase 2|Par-1b
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:88.335 kDa
- Sequence:HGEQLRERRSAAYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Mark1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 600-627 amino acids from the C-terminal region of mouse Mark1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: MARK1 (MAP/microtubule affinity-regulating kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse