Order Entry
Sweden
ContactUsLinkComponent
Anti-SCYL2 Rabbit Polyclonal Antibody
Anti-SCYL2 Rabbit Polyclonal Antibody
Catalog # ABGEAP14165C
Supplier:  Abgent
Anti-SCYL2 Rabbit Polyclonal Antibody
Catalog # ABGEAP14165C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SCY1-like 2
  • Antigen symbol:
    SCYL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    SCY1-like protein 2|KIAA1360|SCYL2|CVAK104|Coated vesicle-associated kinase of 104 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SCYL2 (SCY1-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse