Order Entry
Sweden
ContactUsLinkComponent
Anti-ACTR6 Rabbit Polyclonal Antibody
Anti-ACTR6 Rabbit Polyclonal Antibody
Catalog # ABGEAP13889B
Supplier:  Abgent
undefined
Anti-ACTR6 Rabbit Polyclonal Antibody
Catalog # ABGEAP13889B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ARP6 Actin-Related Protein 6
  • Antigen symbol:
    ACTR6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ACTR6 (ARP6 actin-related protein 6 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human