Anti-VAMP7 Rabbit Polyclonal Antibody
Catalog # ABGEAP13865C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Vesicle-associated membrane protein 7
- Antigen symbol:VAMP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Ti-VAMP|VAMP7|Synaptobrevin-like protein 1|Vesicle-associated membrane protein 7|SYBL1|VAMP-7|Tetanus-insensitive VAMP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: VAMP7 (vesicle-associated membrane protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human