Anti-NOSTRIN Rabbit Polyclonal Antibody
Catalog # ABGEAP13833C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Nitric oxide synthase trafficking
- Antigen symbol:NOSTRIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:BM247 homolog|eNOS-trafficking inducer|Nitric oxide synthase trafficker|Nitric oxide synthase traffic inducer|DaIP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NOSTRIN (nitric oxide synthase trafficking)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse