Anti-UBE2E2 Rabbit Polyclonal Antibody
ABGEAP13824A
: Abgent
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating enzyme E2E 2
- Antigen symbol:UBE2E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:UBCH8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: UBE2E2 (ubiquitin-conjugating enzyme E2E 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human