Anti-H2AFX Rabbit Polyclonal Antibody
Katalog # ABGEAP13716B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:H2A histone family, member X
- Antigen symbol:H2AFX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:H2A histone family member X|H2AFX anitbody|H2A.x|gamma-H2AX|histone H2A|H2AX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human