Anti-TBCC Rabbit Polyclonal Antibody
Catalog # ABGEAP13475A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Tubulin Folding Cofactor C
- Antigen symbol:TBCC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:TBX6|SCDO5|T-box transcription factor TBX6|T-box protein 6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TBCC (tubulin folding cofactor C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse