Anti-MBTPS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP12972A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Membrane-bound transcription factor peptidase site 2
- Antigen symbol:MBTPS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:KFSD|BRESEK|OLMSX|S2P|IFAP|KFSDX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MBTPS2 (membrane-bound transcription factor peptidase, site 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human