Anti-HOXD10 Rabbit Polyclonal Antibody
ABGEAP12608B
: Abgent
- Antibody type:Primary
- Antigen name:Homeobox D10
- Antigen symbol:HOXD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:HOX4D|Hox-4.4|HOX4E|HOX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: HOXD10 (homeobox D10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse